Campylobacter Agar Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C5330 C5331 C5332 C5333
89405-258EA 77.77 USD
89405-258 89405-260 89405-262 89405-264
Campylobacter Agar Base, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Campy Selective Agar Base is used for the selective isolation of Campylobacter jejuni and Campylobacter coli from human, animal, avian and environmental specimens
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Campylobacter Selective Supplement (Karmali), product #SR167E, is required for this medium. Dehydratedculture media with engineered container.
Certifications: BSE Free