Bile Esculin Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C5180 C5181 C5182 C5183
89405-168EA 126.7 USD
89405-168 89405-170 89405-172 89405-174
Bile Esculin Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Bile Esculin Agar (BEA) is recommended for use as a differential medium in the isolation and presumptive identification of enterococci/group D streptococci
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free