Bile Esculin Azide Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
CRITERION™ Bile Esculin Azide Dehydrated Culture Media are recommended for the isolation and differentiation of group D from non-group D streptococci
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
CRITERION™ Dehydratedculture media with engineered container.
Certifications: BSE Free