Brain Heart CC Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C5150 C5151 C5152 C5153
89405-144EA 93.56 USD
89405-144 89405-146 89405-148 89405-150
Brain Heart CC Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Brain Heart CC Agar for the selective isolation of pathogenic fungi from clinical specimens
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Bismuth Sulfite Agar for the selective isolation of Salmonella spp. from clinical and food specimens. Dehydratedculture media with engineered container.
Certifications: BSE Free