Bacillus cereus Medium, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C5060 C5061 C5062 C5063
89405-096EA 104.92 USD
89405-096 89405-098 89405-100 89405-102
Bacillus cereus Medium, CRITERION™, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
CRITERION™ Bacillus Cereus Agar Base is a selective agar medium used for the isolation and determination of Bacillus Cereus
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Egg Yolk Emulsion, product r450290, and Bacillus Cereus Selective Supplement, Polymyxin B, product #SR099E, are required for this medium. Requires supplements: Egg Yolk Suspension 89407-700, and Bacillus Cereus Selective Supplement Polymyxin B SR099E. Dehydratedculture media with engineered container.
Certifications: BSE Free