Azide Dextrose Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C5050
C5051
C5052
C5053
89405-088EA
62.56
USD
89405-088
89405-090
89405-092
89405-094
Azide Dextrose Broth, CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Azide Dextrose Broth is used to detect the presence of streptococci in water, shellfish, sewage, and milk
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free