Crystal violet advanced for Gram staining

Supplier: HARDY DIAGNOSTICS

Synonyms: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet

C128A
89405-046EA 242.82 USD
89405-046
Crystal violet advanced for Gram staining
Crystal Violet

Used as a primary stain in the advanced gram stain procedure


An aqueous solution of crystal violet.

Formula: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
MDL Number: MFCD00011750
CAS Number: 548-62-9
Merck Index: 13,04406

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR