PDX-SIB, Environmental Screen
Supplier: HARDY DIAGNOSTICS
Media for the detection of Salmonella, AOAC RI approved, 24 to 48 h result, closed system, no special equipment needed, use with EnviroMAX sampling sponge on a stick 89405-022.
- Closed system, once the broth media is inoculated the system stays closed
- Three simple steps
The easiest environmental Salmonella test available. This test is both reliable and inexpensive with presumptive results available for the most common Salmonella spp, within 24 to 48 h, no sub-culturing or specimen transfers required.
Just collect your sample, inoculate the PDX-SIB media, and incubate. Sampling devices are not included.
Salmonella selective enrichment media containing a Salmonella specific metabolic indicator. Broth turns yellow when Salmonella species is present. Excellent critical control point surveillance tool.
PDX-SIB was subjected to a panel of more than 100 Salmonella and non-salmonella organisms. The test exhibited 98.5% sensitivity with 95% specificity. The tests were conducted with inocula at both low (<10 CFU per sample) as well as high (>100 CFU per sample).