Heating Mantle, Gallon Can, Glas-Col®
Supplier: Glas-Col
100DDH098BC
89404-900EA 731.98 USD
89404-900
Heating Mantle, Gallon Can, Glas-Col®
Heaters Heating Mantles
Mantle for gallon can.
  • Fabric interior
  • Durable aluminum outer shell
  • Supports weight of gallon can
Provides heat and support for a gallon can and is used speed the removal of viscous materials such as tars, plastics, shortening etc.
Delivery information: 7-10 days after order.