Multi-Pulse Vortexer with Pulsing, Glas-Col
Supplier: Glas-Col
099AVB4
89404-898EA 5404.72 USD
89404-898
Multi-Pulse Vortexer with Pulsing, Glas-Col
Shakers and Mixers Vortex Mixers
The new cartridge design of the Multi Pulse Vortexer allows the user to switch the size of tubes to be mixed by simply changing the cartridge size, thus eliminating the need for extra racks.
  • Interchangeable cartridge system
  • Variable speed 100-2000 rpm
  • Pulse feature interrupts actiion for more thorough mixing
The contents of the test tubes remain visible during mixing and sample can be added without stopping the action.
Ordering information: Each vortexer comes with cartridges for 12-13mm tubes and 15-16mm tubes.
Delivery information: 7-10 days after order.