Epredia™ Cassette Sponge, Epredia
Supplier: Epredia
8453
89404-854CS 77.96 USD
89404-854
Epredia™ Cassette Sponge, Epredia
Biopsy Pads
Protect small biopsies from overprocessing with the Epredia™ Sponge.
  • Blue, open-cell foam sponge holds specimen in cassette