SGE Syringe Syringes for Varian Autosamplers, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
002951
89404-526EA 94.64 USD
89404-526
SGE Syringe Syringes for Varian Autosamplers, Trajan Scientific and Medical
Syringes Syringes for Chromatography
SGE 23 cone autosampler syringes meet all fit, form and function criteria of a specific autosampler model.
  • High injection volume accuracy and reproducibility
  • Designed with submicron tolerances
  • virtually zero dead volume
The extensive range of SGE autosampler syringes meet all fit, form and function criteria of a specific autosampler model. As minimum requirements, they meet dimensional specifications, have accuracy of better than ±2%, and are designed for worry free overnight sampling.