Tygon S3™ B-44-4X Food and Beverage Dispensing Tubing
Supplier: Saint Gobain Life Sciences
AAA00065 AAA00001 AAA00002 AAA00004 AAA00006 AAA00009 AAA00010 AAA00011 AAA00018 AAA00023 AAA00028 AAA00029 AAA00036 AAA00053 AAA00062 AAA00064
89404-196PK 349.74 USD
89404-196 89404-216 89404-224 89404-226 89404-232 89404-238 89404-240 89404-156 89404-158 89404-160 89404-162 89404-164 89404-166 89404-168 89404-170 89404-172 89404-174 89404-176 89404-178 89404-180 89404-182 89404-184 89404-186 89404-188 89404-190 89404-192 89404-194 89404-198 89404-200 89404-202 89404-204 89404-206 89404-208 89404-210 89404-212 89404-214 89404-218 89404-220 89404-222 89404-230 89404-234 89404-236
Tygon S3™ B-44-4X Food and Beverage Dispensing Tubing
Tubing Transfer Tubing
Designed to ensure a bacterial-free fluid path in a wide variety of food processing applications, Tygon S3™ B-44-4X tubing from Saint-Gobain Performance Plastics is now phthalate-free.
Tygon S3™ B-44-4X Food, Milk, and Dairy Tubing features a smooth, non-porous bore that will not trap particulates or promote bacterial growth. Compatible with foods containing a high oil content. Resistant to harsh alkaline cleaners and sanitizers. Excellent alternative to rigid piping systems.
The bio-based Tygon S3™ line combines the high performance standards customers demand with an eco-friendly tubing design. Producers of food, milk, and dairy products insist upon Tygon S3™ B-44-4X for dependable performance in countless filling, draining, transfer, and processing applications. The smooth, non-porous bore inhibits particle entrapment, promoting a sanitary fluid path by minimizing potential for bacteria growth. It offers outstanding resistance to harsh alkaline cleaners and is equally unaffected by commonly used sanitizers. Lightweight and flexible, Tygon S3™ B-44-4X tubing can be put into service quickly. It readily bends to accommodate abrupt corners and obstructions, requiring a minimum of couplings and fittings. Its flexibility can save up to one-third the footage and much of the labor required to install rigid stainless steel or plastic piping. Tygon S3™ B-44-4X tubing is available in large bore sizes (up to 6" inside diameter), making it a flexible alternative to rigid piping systems. It provides complete clarity for positive visual inspection and flow control. A special construction of Tygon S3™ B-44-4X tubing is also available to meet applications involving elevated pressure. In these instances, select Tygon S3™ Pressure Tubing, B-44-4X I.B., which has the identical product features found in Tygon S3™ B-44-4X, with reinforcement added to withstand up to four times the pressure.
Certifications: Tygon S3™ B-44-4X tubing complies with FDA, NSF, 3-A, Japan Food Sanitation Law # 370/1959, REACH, 1935/2004/EC and 10/2011/EU and 10/2011/EU for many foods and beverages. Tygon S3™ tubing products do not contain chemicals listed in California's proposition 65. Contains no BPA or Phthalates.
Ordering information: Tygon S3™ B-44-4X tubing is available in 50', 20', or 10' foot lengths.