Tygon® Laboratory Vacuum Tubing, Saint-Gobain
Supplier: Saint Gobain Life Sciences
ACF00012-V
89403-860PK 216.62 USD
89403-860
Tygon® Laboratory Vacuum Tubing, Saint-Gobain
Tubing Vacuum/Pressure Tubing
The bio-based Tygon® line combines the high performance standards customers demand with an eco-friendly tubing design
Crystal clear and flexible, long-lasting and crack-resistant. Tygon tubing handles most inorganic chemicals found in the laboratory. It is non-oxidizing and non-contaminating, and less permeable than rubber tubing. The glassy-smooth inner bore helps prevent build-up to facilitate cleaning. Coils are marked at 1 foot or 30.4 cm intervals for easy measuring.