VWR® Compound Laboratory Microscopes
Supplier: VWR International
89403-326 89403-328 89403-330 89403-332 89403-342 89403-346 89403-348 89403-350 89403-362 89403-368 89403-370 89403-472 89404-468 89404-470 89496-656 89496-668
89403-326EA 76.66 USD
89403-326 89403-328 89403-330 89403-332 89403-342 89403-346 89403-348 89403-350 89403-362 89403-368 89403-370 89403-472 89404-468 89404-470 89496-656 89496-668
VWR® Compound Laboratory Microscopes
Microscopes
VWR® Compound Laboratory Microscopes, equipped with Planachromatic Objectives, are ideal for laboratory, routine, and research work.
  • Equipped with planachromatic objectives
  • Ergonomic, anti-fatigue design
  • Siedentopf-type head with a 30° inclination and 360° rotating capabilities
  • One-year warranty on electrical parts
  • Five-year warranty on mechanical parts and optics
The series offers an ergonomic, anti- fatigue design to enable long hours of comfortable observation.
This VWR® Compound Microscope features a Siedentopf type head with a 30° inclination and 360° rotating capabilities. Standard features include, widefield eyepieces WF10X / 20mm, reversed quintuple nosepiece, plan objectives PL 4X, 10X, 40X S, 100X S-Oil, coaxial coarse and fine focusing adjustment, built in low position coaxial mechanical stage, centerable and focusable 1.25 N.A Abbe condenser, iris diaphragm with filter holder, Koehler illumination halogen 12V / 20W with intensity control, main supply 110V UL approved.
Certifications: C-UL, UL Listed. CSA certified.
Ordering information: Microscope is supplied with all accessories listed above. The microscope is also supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.