Battery-Powered Benchtop Densitometer Turbidimeter, McFarland Units, DEN-1B, Grant Instruments
Supplier: GRANT USA INC.
DEN-1B
89402-910EA 1487 USD
89402-910
Battery-Powered Benchtop Densitometer Turbidimeter, McFarland Units, DEN-1B, Grant Instruments
Turbidity Meters
The Grant bio DEN-1B is a compact densitometer for measuring turbidity of cell suspensions in a range of life science applications, ideal for measurements on the bench or in cold rooms, incubators and workstations
Small, lightweight, easy to use instruments. Rapid, clear results displayed digitally in McFarland units. Battery operated DEN-1B is convenient for use in remote areas without power. Factory calibrated; retains calibration without power supply. Can be user calibrated with commercial standards or cell suspensions prepared in the laboratory. Light diode light source.
Certifications: CE marked.
Ordering information: Instruments supplied with a D16 adapter for Ø 16 mm tubes. Manufacturers 2 year warranty.