Recirculating Chiller, 1.5 HP, PolyScience
Supplier: Polyscience
6860P46V270D
89400-986EA 14785.21 USD
89400-986
Recirculating Chiller, 1.5 HP, PolyScience
Circulating Thermostats Closed Circulator Systems Cooling Circulators
High capacity cooling for lasers, electron microscopes and other laboratory applications.
Chillers are equipped with large, dual displays, which present temperature and pressure or flow rate simultaneously. The compact, portable design takes up less floor space, without compromising function. These chillers provide cooling at ambient temperatures as high as 35°C.
User-adjustable temperature, pressure, and flow rate alarms.
Simple operation and low-maintenance.
Certifications: CE marked. cTUVus compliant.
Ordering information: 240V/50Hz models are also available; contact your Avantor sales representative for more information.