Miller® Manual Rope Grabs, Honeywell Safety
Supplier: Honeywell Safety Products
8174/U
89400-546EA 182.26 USD
89400-546
Miller® Manual Rope Grabs, Honeywell Safety
Fall Protection Fall Arresters
Manual rope grabs move easily up and down vertical lifelines to provide continuous fall protection.
  • In the event of a fall, rope grabs lock quickly
  • Compact design with corrosion-resistant, stainless steel construction
  • Lightweight and easy to use
  • Designed for use with 16mm or 19mm synthetic rope