Miller® Relief Step Safety Device, Honeywell Safety
Supplier: Honeywell Safety Products
9099X/12-H5
89400-432CS 417.39 USD
89400-432
Miller® Relief Step Safety Device, Honeywell Safety
Fall Protection Fall Lanyards and Ropes
Alleviates the effects of orthostatic intolerance, also known as suspension trauma.
  • Easy to deploy and use
  • Attaches to any brand full-body harness
  • Provides support and enhances blood circulation until rescue, permitting the ability to move and flex leg muscles