HyClone™ HyCryo Media 2x Cryopreservation Media, HyClone products (Cytiva)
Supplier: Cytiva
SR30001.02
89399-932EA 223.87 USD
89399-932
HyClone™ HyCryo Media 2x Cryopreservation Media, HyClone products (Cytiva)
Cell Freezing Media
HyCryo Media - 2x cryopreservation media for general use
Animal Origin free Serum free media is a serum-free, animal origin-free product intended for cryogenic storage for your standard cell lines.