Anti-PDXP Rabbit Polyclonal Antibody

Supplier: Abgent
AP9822A
89399-184EA 589.74 USD
89399-184
Anti-PDXP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDXP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR