Anti-CHMP4B Rabbit Polyclonal Antibody

Supplier: Abgent
AP9528A
89398-588EA 619.55 USD
89398-588
Anti-CHMP4B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CHMP4B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR