Anti-ANP32A Rabbit Polyclonal Antibody

Supplier: Abgent
AP9520B
89398-572EA 619.21 USD
89398-572
Anti-ANP32A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ANP32A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR