Anti-CNTN1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP9490C
89398-512EA 619.21 USD
89398-512
Anti-CNTN1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CNTN1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR