Anti-CD209 Rabbit Polyclonal Antibody

Supplier: Abgent
AP9456C
89398-446EA 619.21 USD
89398-446
Anti-CD209 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD209
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR