Anti-NPY5R Rabbit Polyclonal Antibody

Supplier: Abgent
AP9265C
89398-052EA 589.56 USD
89398-052
Anti-NPY5R Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NPY5R
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR