Anti-TOP2A Rabbit Polyclonal Antibody

Supplier: Abgent
AP9248B
89398-016EA 619.55 USD
89398-016
Anti-TOP2A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TOP2A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR