Anti-SLC22A6 Rabbit Polyclonal Antibody

Supplier: Abgent
AP9237B
89397-994EA 619.21 USD
89397-994
Anti-SLC22A6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLC22A6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR