Anti-C19orf50 Rabbit Polyclonal Antibody

Supplier: Abgent
AP9140D
89397-796EA 619.21 USD
89397-796
Anti-C19orf50 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: C19orf50
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR