Anti-CDK19 Rabbit Polyclonal Antibody

Supplier: Abgent
AP9082C
89397-676EA 619.21 USD
89397-676
Anti-CDK19 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDK19
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR