Anti-CNKSR3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP8957B
89397-434EA 589.74 USD
89397-434
Anti-CNKSR3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CNKSR3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR