Anti-ALOX12 Rabbit Polyclonal Antibody

Supplier: Abgent
AP8877B
89397-276EA 619.55 USD
89397-276
Anti-ALOX12 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ALOX12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR