Anti-SLC22A5 Rabbit Polyclonal Antibody

Supplier: Abgent
AP8854C
89397-226EA 589.74 USD
89397-226
Anti-SLC22A5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLC22A5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR