Anti-ACPT Rabbit Polyclonal Antibody

Supplier: Abgent
AP8714A
89396-948EA 589.74 USD
89396-948
Anti-ACPT Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ACPT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR