Anti-COMMD3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP8640C
89396-788EA 589.74 USD
89396-788
Anti-COMMD3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: COMMD3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR