Anti-MLX Rabbit Polyclonal Antibody

Supplier: Abgent
AP8638C
89396-784EA 619.21 USD
89396-784
Anti-MLX Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MLX
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR