Anti-CYP4Z1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7886A
89395-488EA 619.55 USD
89395-488
Anti-CYP4Z1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CYP4Z1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR