Anti-CRK Rabbit Polyclonal Antibody

Supplier: Abgent
AP7831A
89395-316EA 589.74 USD
89395-316
Anti-CRK Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CRK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR