Anti-CSF1R Rabbit Polyclonal Antibody

Supplier: Abgent
AP7604D
89394-538EA 619.21 USD
89394-538
Anti-CSF1R Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CSF1R
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR