Anti-PI4K2A Rabbit Polyclonal Antibody

Supplier: Abgent
AP7575D
89394-450EA 589.74 USD
89394-450
Anti-PI4K2A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PI4K2A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR