Anti-RAD50 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7360A
89393-806EA 589.74 USD
89393-806
Anti-RAD50 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RAD50
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR