Anti-MME Rabbit Polyclonal Antibody

Supplier: Abgent
AP7329B
89393-716EA 619.21 USD
89393-716
Anti-MME Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MME
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR