Anti-MEOX1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7267A
89393-526EA 619.55 USD
89393-526
Anti-MEOX1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MEOX1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR