Anti-KSR1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7202A
89393-306EA 619.04 USD
89393-306
Anti-KSR1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KSR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR