Anti-PDPK1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7042F
89392-824EA 589.74 USD
89392-824
Anti-PDPK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDPK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR