Anti-ST8SIA2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6917A
89392-492EA 589.74 USD
89392-492
Anti-ST8SIA2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ST8SIA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR