Anti-RPS18 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6887C
89392-428EA 619.04 USD
89392-428
Anti-RPS18 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RPS18
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR