Anti-IGF2BP3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6763C
89392-100EA 589.74 USD
89392-100
Anti-IGF2BP3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IGF2BP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR