Anti-GBAS Rabbit Polyclonal Antibody

Supplier: Abgent
AP6752C
89392-076EA 619.55 USD
89392-076
Anti-GBAS Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR