Anti-TYMS Rabbit Polyclonal Antibody

Supplier: Abgent
AP6682B
89391-878EA 615.89 USD
89391-878
Anti-TYMS Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TYMS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR